Order Entry
Export
ContactUsLinkComponent
3,5-Dimethoxypicolinonitrile 95%
3,5-Dimethoxypicolinonitrile 95%
Catalog # APOSOR950635-1G
Supplier:  Apollo Scientific
CAS Number:  
3,5-Dimethoxypicolinonitrile 95%
Catalog # APOSOR950635-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.161 g/mol
Storage Temperature:Ambient
CAS Number:36057-45-1
Formula:C₈H₈N₂O₂
MDL Number:MFCD18255817