Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ethyl 4,5-dimethyl-1H-pyrrole-2-carboxylate
Ethyl 4,5-dimethyl-1H-pyrrole-2-carboxylate 95%
Ethyl 4,5-dimethyl-1H-pyrrole-2-carboxylate 95%
Ethyl 4,5-dimethyl-1H-pyrrole-2-carboxylate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Ethyl 4,5-dimethyl-1H-pyrrole-2-carboxylate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
2199-45-3
:
Formula:
C₉H₁₃NO₂
MDL Number:
MFCD04112537
:
167.205 g/mol
Storage Temperature:
Ambient