0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
6-Azaspiro[3.4]octan-2-ylmethanol
6-Azaspiro[3.4]octan-2-ylmethanol
6-Azaspiro[3.4]octan-2-ylmethanol
6-Azaspiro[3.4]octan-2-ylmethanol
Supplier:
Apollo Scientific
Certificates
Expand image
6-Azaspiro[3.4]octan-2-ylmethanol
Supplier:
Apollo Scientific
About this item
CAS Number:
1420958-53-7
:
Formula:
C₈H₁₅NO
MDL Number:
MFCD24384534
:
141.211 g/mol
Storage Temperature:
Ambient