Spiro[2.3]hexan-4-one
Catalog # APOSOR948321-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:96.127 g/mol
Storage Temperature:Ambient
CAS Number:20571-15-7
Formula:C₆H₈O
MDL Number:MFCD00604224