Order Entry
Export
ContactUsLinkComponent
5-Ethoxy-1H-pyrrolo[3,2-b]pyridine 95%
5-Ethoxy-1H-pyrrolo[3,2-b]pyridine 95%
Catalog # APOSOR947697-1G
Supplier:  Apollo Scientific
CAS Number:  
5-Ethoxy-1H-pyrrolo[3,2-b]pyridine 95%
Catalog # APOSOR947697-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :162.189 g/mol
Storage Temperature:Ambient
CAS Number:23612-31-9
Formula:C₉H₁₀N₂O
MDL Number:MFCD13178620