Order Entry
Export
ContactUsLinkComponent
2,6-Diazaspiro[3.4]octan-7-one 95%
Irritant
2,6-Diazaspiro[3.4]octan-7-one 95%
Catalog # APOSOR947562-250MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
2,6-Diazaspiro[3.4]octan-7-one 95%
Catalog # APOSOR947562-250MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

About this item

 :
 :126.156 g/mol
 :GHS07
Storage Temperature:Refrigerator
CAS Number:1211515-65-9
Formula:C₆H₁₀N₂O
MDL Number:MFCD21364482