Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,4-Dimethyl-5-nitroaniline
3,4-Dimethyl-5-nitroaniline 95%
3,4-Dimethyl-5-nitroaniline 95%
3,4-Dimethyl-5-nitroaniline 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3,4-Dimethyl-5-nitroaniline 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
64823-22-9
:
Formula:
C₈H₁₀N₂O₂
MDL Number:
MFCD09880021
:
166.177 g/mol
Storage Temperature:
Ambient