Order Entry
Export
ContactUsLinkComponent
N,N'-(1,3-Phenylene)diacetamide 95%
N,N'-(1,3-Phenylene)diacetamide 95%
Catalog # APOSOR947264-1G
Supplier:  Apollo Scientific
CAS Number:  
N,N'-(1,3-Phenylene)diacetamide 95%
Catalog # APOSOR947264-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :192.214 g/mol
Storage Temperature:Ambient
CAS Number:10268-78-7
Formula:C₁₀H₁₂N₂O₂
MDL Number:MFCD00035792