Order Entry
Export
ContactUsLinkComponent
N1,N2-Diethylbenzene-1,2-diamine 95%
N1,N2-Diethylbenzene-1,2-diamine 95%
Catalog # APOSOR947185-1G
Supplier:  Apollo Scientific
CAS Number:  
N1,N2-Diethylbenzene-1,2-diamine 95%
Catalog # APOSOR947185-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :164.247 g/mol
Storage Temperature:Ambient
CAS Number:24340-87-2
Formula:C₁₀H₁₆N₂
MDL Number:MFCD14582999