Order Entry
Export
ContactUsLinkComponent
5-(2-Chloroethyl)-2,3-dihydrobenzofuran 95%
5-(2-Chloroethyl)-2,3-dihydrobenzofuran 95%
Catalog # APOSOR946504-1G
Supplier:  Apollo Scientific
CAS Number:  
5-(2-Chloroethyl)-2,3-dihydrobenzofuran 95%
Catalog # APOSOR946504-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :182.647 g/mol
Storage Temperature:Ambient
CAS Number:943034-50-2
Formula:C₁₀H₁₁ClO
MDL Number:MFCD11046521