Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
2-Chloro-5,7-dihydrofuro[3,4-b]pyridine
2-Chloro-5,7-dihydrofuro[3,4-b]pyridine ≥98%
2-Chloro-5,7-dihydrofuro[3,4-b]pyridine ≥98%
Irritant
2-Chloro-5,7-dihydrofuro[3,4-b]pyridine ≥98%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
2-Chloro-5,7-dihydrofuro[3,4-b]pyridine ≥98%
Supplier:
Apollo Scientific
About this item
CAS Number:
1464091-43-7
:
Formula:
C₇H₆ClNO
MDL Number:
MFCD24386279
:
155.582 g/mol
:
GHS07
Storage Temperature:
Refrigerator