Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chlorothiazolo[5,4-b]pyridine
5-Chlorothiazolo[5,4-b]pyridine 99%
5-Chlorothiazolo[5,4-b]pyridine 99%
Irritant
5-Chlorothiazolo[5,4-b]pyridine 99%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-Chlorothiazolo[5,4-b]pyridine 99%
Supplier:
Apollo Scientific
About this item
CAS Number:
1313726-12-3
:
Formula:
C₆H₃ClN₂S
MDL Number:
MFCD22199690
:
170.619 g/mol
:
GHS07
Storage Temperature:
Ambient