
6-(Ethylsulphonyl)-2H-1,4-benzoxazin-3(4H)-one
Catalog # APOSOR9456-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:241.264 g/mol
:GHS07
Storage Temperature:Ambient
CAS Number:874774-43-3
Formula:C₁₀H₁₁NO₄S
MDL Number:MFCD00798368