
1,2-Benzoxathiin-4(3H)-one 2,2-dioxide
Catalog # APOSOR9453-100MG
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:198.196 g/mol
Melting Pt:88…92 °C
:GHS07
Storage Temperature:Ambient
CAS Number:49670-47-5
Formula:C₈H₆O₄S
MDL Number:MFCD00798366