Order Entry
Export
ContactUsLinkComponent
4-Amino-2,6-diethylbenzonitrile 95%
4-Amino-2,6-diethylbenzonitrile 95%
Catalog # APOSOR942650-1G
Supplier:  Apollo Scientific
CAS Number:  
4-Amino-2,6-diethylbenzonitrile 95%
Catalog # APOSOR942650-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :174.242 g/mol
Storage Temperature:Ambient
CAS Number:1003708-27-7
Formula:C₁₁H₁₄N₂
MDL Number:MFCD08144071