2,6-Dichloronicotinamide 95%
Supplier: Apollo Scientific
Jump to:
About this item
Synonyms: 2,6-Dichloropyridine-3-carboxamide
- CAS Number:62068-78-4
- Formula:C₆H₄Cl₂N₂O
- MDL Number:MFCD03230010
- Molecular Weight:191.015 g/mol
- Storage Temperature:Ambient
6 Options Available Below
Product Details & Documents
Synonyms: 2,6-Dichloropyridine-3-carboxamide
- CAS Number:62068-78-4
- Formula:C₆H₄Cl₂N₂O
- MDL Number:MFCD03230010
- Molecular Weight:191.015 g/mol
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product
- Specifications:New ProductNew Product



