Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5,6-Dichloropyridin-2-amine
5,6-Dichloropyridin-2-amine 95%
5,6-Dichloropyridin-2-amine 95%
5,6-Dichloropyridin-2-amine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
5,6-Dichloropyridin-2-amine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1192814-45-1
:
Formula:
C₅H₄Cl₂N₂
MDL Number:
MFCD17012012
:
163.005 g/mol
Storage Temperature:
Ambient