Order Entry
Export
ContactUsLinkComponent
 
3-Hydroxy-2,2-dimethylpropanal 95%
Catalog # APOSOR941564-1G
Supplier:  Apollo Scientific
CAS Number:  
3-Hydroxy-2,2-dimethylpropanal 95%
Catalog # APOSOR941564-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :102.132 g/mol
Storage Temperature:Ambient
CAS Number:597-31-9
Formula:C₅H₁₀O₂
MDL Number:MFCD00006963