Order Entry
Export
ContactUsLinkComponent
Methyl 2-methyl-3-oxopentanoate 95%
Methyl 2-methyl-3-oxopentanoate 95%
Catalog # APOSOR941458-1G
Supplier:  Apollo Scientific
CAS Number:  
Methyl 2-methyl-3-oxopentanoate 95%
Catalog # APOSOR941458-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :144.168 g/mol
Storage Temperature:Ambient
CAS Number:17422-12-7
Formula:C₇H₁₂O₃
MDL Number:MFCD00184703