5-Oxa-8-azaspiro[3.5]nonane
Catalog # APOSOR940852-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:127.184 g/mol
Storage Temperature:Ambient
CAS Number:220291-93-0
Formula:C₇H₁₃NO
MDL Number:MFCD18382670