Order Entry
Export
ContactUsLinkComponent
7-Methyl[1,2,4]triazolo[1,5-a]pyridine 95%
7-Methyl[1,2,4]triazolo[1,5-a]pyridine 95%
Catalog # APOSOR940566-1G
Supplier:  Apollo Scientific
CAS Number:  
7-Methyl[1,2,4]triazolo[1,5-a]pyridine 95%
Catalog # APOSOR940566-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :133.151 g/mol
Storage Temperature:Ambient
CAS Number:4999-42-2
Formula:C₇H₇N₃
MDL Number:MFCD10699196