Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-Amino-2,3-dichlorophenol
4-Amino-2,3-dichlorophenol 97%
4-Amino-2,3-dichlorophenol 97%
Irritant
4-Amino-2,3-dichlorophenol 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
4-Amino-2,3-dichlorophenol 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
39183-17-0
:
Formula:
C₆H₅Cl₂NO
MDL Number:
MFCD14455913
:
178.016 g/mol
:
GHS07
Storage Temperature:
Ambient