0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
6,10-Dioxaspiro[4.5]decane-7,9-dione
6,10-Dioxaspiro[4.5]decane-7,9-dione 95%
6,10-Dioxaspiro[4.5]decane-7,9-dione 95%
6,10-Dioxaspiro[4.5]decane-7,9-dione 95%
Supplier:
Apollo Scientific
Certificates
Expand image
6,10-Dioxaspiro[4.5]decane-7,9-dione 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
58093-05-3
:
Formula:
C₈H₁₀O₄
MDL Number:
MFCD01617888
:
170.163 g/mol
Storage Temperature:
Ambient