Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Dimethyl 2,2-dimethylmalonate
Dimethyl 2,2-dimethylmalonate 95%
Dimethyl 2,2-dimethylmalonate 95%
Dimethyl 2,2-dimethylmalonate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Dimethyl 2,2-dimethylmalonate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
6065-54-9
:
Formula:
C₇H₁₂O₄
MDL Number:
MFCD09833516
:
160.168 g/mol
Storage Temperature:
Ambient