Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Dichloro-2,2-dimethoxypropane
1,3-Dichloro-2,2-dimethoxypropane 95%
1,3-Dichloro-2,2-dimethoxypropane 95%
1,3-Dichloro-2,2-dimethoxypropane 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1,3-Dichloro-2,2-dimethoxypropane 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
6626-57-9
:
Formula:
C₅H₁₀Cl₂O₂
MDL Number:
MFCD00130264
:
173.038 g/mol
Storage Temperature:
Ambient