Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-2,2-dimethylpropionic acid
3-Bromo-2,2-dimethylpropionic acid 95%
3-Bromo-2,2-dimethylpropionic acid 95%
3-Bromo-2,2-dimethylpropionic acid 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Bromo-2,2-dimethylpropionic acid 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
2843-17-6
:
Formula:
C₅H₉BrO₂
MDL Number:
MFCD00055490
:
181.028 g/mol
Storage Temperature:
Ambient
UN:
3261
ADR:
8,III