Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1H-Pyrrolo[3,2-c]pyridin-4-amine
1H-Pyrrolo[3,2-c]pyridin-4-amine 97%
1H-Pyrrolo[3,2-c]pyridin-4-amine 97%
APOSOR938784-250MG
:
Apollo Scientific
:
1H-Pyrrolo[3,2-c]pyridin-4-amine 97%
APOSOR938784-250MG
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
250 mg
Seq:
2590
:
:
133.151 g/mol
:
GHS07
:
Refrigerator
:
60290-23-5
:
C₇H₇N₃
:
MFCD08690132