Order Entry
Export
Orders LinkContactUsLinkComponent
(S)-1-(3,4-Dimethoxyphenyl)ethylamine 95%

(S)-1-(3,4-Dimethoxyphenyl)ethylamine 95%

Bottle, 1 g
Catalog # APOSOR938552-1G
Supplier:   Apollo Scientific
(S)-1-(3,4-Dimethoxyphenyl)ethylamine 95%
Catalog # APOSOR938552-1G
Supplier:  Apollo Scientific
CAS Number:  65451-89-0
(S)-1-(3,4-Dimethoxyphenyl)ethylamine 95%

Bottle, 1 g

CAS Number:  65451-89-0
Price
150,00 €
Log in for availability
1 * 1 g
Product Details & Documents
Synonyms:  (S)-1-(3,4-Dimethoxyphenyl)ethanamine
  • CAS Number:65451-89-0
  • Formula:C₁₀H₁₅NO₂
  • MDL Number:MFCD06761896
  • Molecular Weight:181.232 g/mol
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g

Specifications