Order Entry
Export
ContactUsLinkComponent
2,2-Dimethyl-piperazine dihydrochloride 95%
2,2-Dimethyl-piperazine dihydrochloride 95%
Catalog # APOSOR932791-1G
Supplier:  Apollo Scientific
CAS Number:  
2,2-Dimethyl-piperazine dihydrochloride 95%
Catalog # APOSOR932791-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :187.111 g/mol
Storage Temperature:Ambient
CAS Number:128427-07-6
Formula:C₆H₁₄N₂·2ClH
MDL Number:MFCD08752613