Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-Chloro-2-hydroxy-4-methoxy-benzaldehyde
5-Chloro-2-hydroxy-4-methoxy-benzaldehyde 95%
5-Chloro-2-hydroxy-4-methoxy-benzaldehyde 95%
5-Chloro-2-hydroxy-4-methoxy-benzaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
5-Chloro-2-hydroxy-4-methoxy-benzaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
89938-56-7
:
Formula:
C₈H₇ClO₃
MDL Number:
MFCD08059080
:
186.592 g/mol
Storage Temperature:
Ambient