0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
2,4-Dichloro-5-methylphenol
2,4-Dichloro-5-methylphenol 95%
2,4-Dichloro-5-methylphenol 95%
2,4-Dichloro-5-methylphenol 95%
Supplier:
Apollo Scientific
Certificates
Expand image
2,4-Dichloro-5-methylphenol 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1124-07-8
:
Formula:
C₇H₆Cl₂O
MDL Number:
MFCD11520109
:
177.028 g/mol
Melting Pt:
73 °C
Storage Temperature:
Ambient