Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,4-Dimethoxy-2,3-dimethylbenzene
1,4-Dimethoxy-2,3-dimethylbenzene 95%
1,4-Dimethoxy-2,3-dimethylbenzene 95%
1,4-Dimethoxy-2,3-dimethylbenzene 95%
APOSOR931246-1G
:
Apollo Scientific
:
1,4-Dimethoxy-2,3-dimethylbenzene 95%
APOSOR931246-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
166.217 g/mol
:
Ambient
:
39021-83-5
:
C₁₀H₁₄O₂
:
MFCD00094721