Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Trimethylsilyl)benzaldehyde
4-(Trimethylsilyl)benzaldehyde 95%
4-(Trimethylsilyl)benzaldehyde 95%
4-(Trimethylsilyl)benzaldehyde 95%
Supplier:
Apollo Scientific
Certificates
Expand image
4-(Trimethylsilyl)benzaldehyde 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
2199-32-8
:
Formula:
C₁₀H₁₄OSi
MDL Number:
MFCD08064053
:
178.303 g/mol
Storage Temperature:
Ambient