Order Entry
Export
Orders LinkContactUsLinkComponent
2,3'-Bipyridine 95%

2,3'-Bipyridine 95%

Supplier:   Apollo Scientific
2,3'-Bipyridine 95%
Supplier:  Apollo Scientific
Certificates
2,3'-Bipyridine 95%

5 Options Available Below

Product Details & Documents
  • CAS Number:581-50-0
  • Formula:C₁₀H₈N₂
  • MDL Number:MFCD00006215
  • Molecular Weight:156.184 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 21,80 €
    21,80 €
    1 * 1 g
       21,80 €

  • Bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 29,00 €
    29,00 €
    1 * 5 g
       29,00 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 109,00 €
    109,00 €
    1 * 25 g
       109,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - 433,00 €
    433,00 €
    1 * 100 g
       433,00 €

    Specifications:
    New Product
    New Product
  • Bottle
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - 2 150,00 €
    2 150,00 €
    1 * 500 g
       2 150,00 €

    Specifications:
    New Product
    New Product