Order Entry
Export
ContactUsLinkComponent
2,4-Dichloro-6-(methylthio)-1,3,5-triazine 95%
2,4-Dichloro-6-(methylthio)-1,3,5-triazine 95%
Catalog # APOSOR930794-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Dichloro-6-(methylthio)-1,3,5-triazine 95%
Catalog # APOSOR930794-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :196.058 g/mol
Storage Temperature:Ambient
CAS Number:13705-05-0
Formula:C₄H₃Cl₂N₃S
MDL Number:MFCD01679001