Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3-Bromo-4,5-dimethylpyridine
3-Bromo-4,5-dimethylpyridine 97%
3-Bromo-4,5-dimethylpyridine 97%
3-Bromo-4,5-dimethylpyridine 97%
Supplier:
Apollo Scientific
Certificates
Expand image
3-Bromo-4,5-dimethylpyridine 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
27063-98-5
:
Formula:
C₇H₈BrN
MDL Number:
MFCD11656269
:
186.049 g/mol
Storage Temperature:
Ambient