Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,5-Dichloropyrazine-2-carboxamide
3,5-Dichloropyrazine-2-carboxamide 98%
3,5-Dichloropyrazine-2-carboxamide 98%
3,5-Dichloropyrazine-2-carboxamide 98%
Supplier:
Apollo Scientific
Certificates
Expand image
3,5-Dichloropyrazine-2-carboxamide 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
312736-50-8
:
Formula:
C₅H₃Cl₂N₃O
MDL Number:
MFCD10000837
:
192.003 g/mol
Storage Temperature:
Ambient