Order Entry
Export
ContactUsLinkComponent
2,6-Dimethyl-1H-indole 95%
2,6-Dimethyl-1H-indole 95%
Catalog # APOSOR929350-1G
Supplier:  Apollo Scientific
CAS Number:  
2,6-Dimethyl-1H-indole 95%
Catalog # APOSOR929350-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :145.201 g/mol
Storage Temperature:Ambient
CAS Number:5649-36-5
Formula:C₁₀H₁₁N
MDL Number:MFCD04117975