Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Methyl 4-methyl-1H-pyrrole-3-carboxylate
Methyl 4-methyl-1H-pyrrole-3-carboxylate 95%
Methyl 4-methyl-1H-pyrrole-3-carboxylate 95%
Methyl 4-methyl-1H-pyrrole-3-carboxylate 95%
Supplier:
Apollo Scientific
Certificates
Expand image
Methyl 4-methyl-1H-pyrrole-3-carboxylate 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
40318-15-8
:
Formula:
C₇H₉NO₂
MDL Number:
MFCD10565693
:
139.152 g/mol
Storage Temperature:
Ambient