Order Entry
Export
ContactUsLinkComponent
1,5-Dihydro-benzo[e][1,4]oxazepin-2-one 95%
1,5-Dihydro-benzo[e][1,4]oxazepin-2-one 95%
Catalog # APOSOR928769-1G
Supplier:  Apollo Scientific
CAS Number:  
1,5-Dihydro-benzo[e][1,4]oxazepin-2-one 95%
Catalog # APOSOR928769-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :163.173 g/mol
Storage Temperature:Ambient
CAS Number:3693-08-1
Formula:C₉H₉NO₂
MDL Number:MFCD11041283