Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,6-Diethylpyridine 95%

Supplier:   Apollo Scientific
2,6-Diethylpyridine 95%
Supplier:  Apollo Scientific
Certificates
2,6-Diethylpyridine 95%

2 Options Available Below

Product Details & Documents
  • CAS Number:935-28-4
  • Formula:C₉H₁₃N
  • MDL Number:MFCD00049215
  • Molecular Weight:135.206 g/mol
  • Storage Temperature:Ambient
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Bottle
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 34,80 €
    34,80 €
    1 * 1 g
       34,80 €

  • Bottle
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 168,00 €
    168,00 €
    1 * 5 g
       168,00 €

    Specifications:
    New Product
    New Product