Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-Boc-4-(hydroxymethyl)pyrazole
1-Boc-4-(hydroxymethyl)pyrazole 98%
1-Boc-4-(hydroxymethyl)pyrazole 98%
1-Boc-4-(hydroxymethyl)pyrazole 98%
Supplier:
Apollo Scientific
Certificates
Expand image
1-Boc-4-(hydroxymethyl)pyrazole 98%
Supplier:
Apollo Scientific
About this item
CAS Number:
199003-22-0
:
Formula:
C₉H₁₄N₂O₃
MDL Number:
MFCD24335255
:
198.219 g/mol
Storage Temperature:
Ambient