Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Hydroxymethyl)-5-methyl-1,3-dioxol-2-one
4-(Hydroxymethyl)-5-methyl-1,3-dioxol-2-one 96%
4-(Hydroxymethyl)-5-methyl-1,3-dioxol-2-one 96%
4-(Hydroxymethyl)-5-methyl-1,3-dioxol-2-one 96%
Supplier:
Apollo Scientific
Certificates
Expand image
4-(Hydroxymethyl)-5-methyl-1,3-dioxol-2-one 96%
Supplier:
Apollo Scientific
About this item
CAS Number:
91526-18-0
:
Formula:
C₅H₆O₄
MDL Number:
MFCD12165896
:
130.099 g/mol
Boiling Pt:
214 °C
Storage Temperature:
Freezer