Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
N-Methyl-4-phenoxybenzylamine
N-Methyl-4-phenoxybenzylamine
N-Methyl-4-phenoxybenzylamine
APOSOR9217-1G
:
Apollo Scientific
:
N-Methyl-4-phenoxybenzylamine
APOSOR9217-1G
:
Apollo Scientific
:
Pack type:
Bottle
Pk:
1 g
Seq:
3047
:
:
213.275 g/mol
:
Ambient
:
169943-40-2
:
C₁₄H₁₅NO
:
MFCD02089415