7-Benzyl-2,7-diazaspiro[4.4]nonan-3-one
Catalog # APOSOR921245-1G
Supplier: Apollo Scientific
CAS Number:
Specifications
- Pack type:Bottle
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:230.306 g/mol
Storage Temperature:Ambient
CAS Number:909723-04-2
Formula:C₁₄H₁₈N₂O
MDL Number:MFCD11043248