Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1-(2,4-Dimethylpyrimidin-5-yl)ethan-1-one
1-(2,4-Dimethylpyrimidin-5-yl)ethan-1-one 95%
1-(2,4-Dimethylpyrimidin-5-yl)ethan-1-one 95%
1-(2,4-Dimethylpyrimidin-5-yl)ethan-1-one 95%
Supplier:
Apollo Scientific
Certificates
Expand image
1-(2,4-Dimethylpyrimidin-5-yl)ethan-1-one 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
55933-85-2
:
Formula:
C₈H₁₀N₂O
MDL Number:
MFCD00053574
:
150.178 g/mol
Storage Temperature:
Ambient