Order Entry
Export
ContactUsLinkComponent
 
2,4-Dichloro-3-hydroxybenzaldehyde 95%
Catalog # APOSOR920426-1G
Supplier:  Apollo Scientific
CAS Number:  
2,4-Dichloro-3-hydroxybenzaldehyde 95%
Catalog # APOSOR920426-1G
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :191.011 g/mol
Storage Temperature:Ambient
CAS Number:56962-13-1
Formula:C₇H₄Cl₂O₂
MDL Number:MFCD22199563