Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
5-(Isocyanatomethyl)-1,3-benzodioxole
5-(Isocyanatomethyl)-1,3-benzodioxole 97%
5-(Isocyanatomethyl)-1,3-benzodioxole 97%
Irritant
5-(Isocyanatomethyl)-1,3-benzodioxole 97%
Supplier:
Apollo Scientific
Certificates
Expand image
Irritant
5-(Isocyanatomethyl)-1,3-benzodioxole 97%
Supplier:
Apollo Scientific
About this item
CAS Number:
71217-46-4
:
Formula:
C₉H₇NO₃
MDL Number:
MFCD08435909
:
177.157 g/mol
Storage Temperature:
Refrigerator
UN:
2206
ADR:
6.1,III
:
GHS07