Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
6-Methoxy-1H-pyrrolo[3,2-c]pyridine
6-Methoxy-1H-pyrrolo[3,2-c]pyridine 95%
6-Methoxy-1H-pyrrolo[3,2-c]pyridine 95%
6-Methoxy-1H-pyrrolo[3,2-c]pyridine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
6-Methoxy-1H-pyrrolo[3,2-c]pyridine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
80862-08-4
:
Formula:
C₈H₈N₂O
MDL Number:
MFCD08690135
:
148.162 g/mol
Storage Temperature:
Ambient