Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
3,6-Dimethoxy-4-hydroxypyridazine
3,6-Dimethoxy-4-hydroxypyridazine 95%
3,6-Dimethoxy-4-hydroxypyridazine 95%
3,6-Dimethoxy-4-hydroxypyridazine 95%
Supplier:
Apollo Scientific
Certificates
Expand image
3,6-Dimethoxy-4-hydroxypyridazine 95%
Supplier:
Apollo Scientific
About this item
CAS Number:
1428532-82-4
:
Formula:
C₆H₈N₂O₃
MDL Number:
MFCD23701100
:
156.139 g/mol
Storage Temperature:
Ambient